Assay Details
| Gene Symbol: LOC4334137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775042.2 | XP_015630528.1 | 1 - 2 | 1593 | 79 | |
| GenBank mRNA | AK069303.1 | - | 1 - 2 | 343 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015775042.2 transcripts.
Set Membership: |
|

LOC4334137
Genomic Map