Assay Details
| Gene Symbol: LOC4334137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775042.2 | XP_015630528.1 | 2 - 3 | 2528 | 69 | |
| XM_015775043.2 | XP_015630529.1 | 2 - 3 | 1819 | 69 | ||
| XM_015775044.2 | XP_015630530.1 | 2 - 3 | 2505 | 69 | ||
| XM_015775045.2 | XP_015630531.1 | 2 - 3 | 1796 | 69 | ||
| GenBank mRNA | AK069303.1 | - | 2 - 3 | 1278 | 69 | |
| AK289008.1 | - | 1906 | 69 | |||
More Information
Set Membership: |
|

LOC4334137
Genomic Map