Assay Details
| Gene Symbol: LOC4334141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775549.2 | XP_015631035.1 | 3 - 4 | 728 | 66 | |
| GenBank mRNA | AB003324.1 | - | 3 - 4 | 587 | 66 | |
| AK100227.1 | - | 3 - 4 | 612 | 66 | ||
| AY551917.1 | - | 3 - 4 | 429 | 66 | ||
| AY551922.1 | - | 3 - 4 | 447 | 66 | ||
| AY551926.1 | - | 3 - 4 | 479 | 66 | ||
More Information
Set Membership: |
|

LOC4334141
Genomic Map