Assay Details
| Gene Symbol: LOC4334141 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775549.2 | XP_015631035.1 | 6 - 7 | 909 | 71 | |
| GenBank mRNA | AB003324.1 | - | 6 - 7 | 768 | 71 | |
| AK100227.1 | - | 6 - 7 | 793 | 71 | ||
| AY551917.1 | - | 6 - 7 | 610 | 71 | ||
| AY551922.1 | - | 6 - 7 | 628 | 71 | ||
More Information
Set Membership: |
|

LOC4334141
Genomic Map