Assay Details
| Gene Symbol: LOC4334311 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775601.2 | XP_015631087.1 | 1 - 2 | 202 | 94 | |
| GenBank mRNA | AK059874.1 | - | 1 - 2 | 98 | 94 | |
| AK103085.1 | - | 1 - 2 | 199 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015775601.2 transcripts.
Set Membership: |
|

LOC4334311
Genomic Map