Assay Details
| Gene Symbol: LOC4334311 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775601.2 | XP_015631087.1 | 3 - 4 | 367 | 86 | |
| XM_015775602.2 | XP_015631088.1 | 3 - 4 | 348 | 86 | ||
| GenBank mRNA | AK059874.1 | - | 3 - 4 | 263 | 86 | |
| AK103085.1 | - | 3 - 4 | 364 | 86 | ||
More Information
Set Membership: |
|

LOC4334311
Genomic Map