Assay Details
| Gene Symbol: LOC4334754 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774208.2 | XP_015629694.1 | 7 - 8 | 1351 | 98 | |
| GenBank mRNA | AK068101.1 | - | 7 - 8 | 1361 | 98 | |
More Information
Set Membership: |
|

LOC4334754
Genomic Map