Assay Details
| Gene Symbol: LOC4335013 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778627.2 | XP_015634113.1 | 3 - 4 | 388 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4335013| Gene Symbol: LOC4335013 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778627.2 | XP_015634113.1 | 3 - 4 | 388 | 70 | |
Set Membership: |
|