Assay Details
| Gene Symbol: LOC4335013 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778627.2 | XP_015634113.1 | 6 - 7 | 1266 | 81 | |
| GenBank mRNA | AK060609.1 | - | 88 | 81 | ||
More Information
Set Membership: |
|

LOC4335013
Genomic Map