Assay Details
| Gene Symbol: LOC4335094 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778097.2 | XP_015633583.1 | 2 - 3 | 369 | 98 | |
| GenBank mRNA | AB126934.1 | - | 2 - 3 | 289 | 98 | |
| AK119327.1 | - | 2 - 3 | 348 | 98 | ||
| AY347880.1 | - | 2 - 3 | 289 | 98 | ||
More Information
Set Membership: |
|

LOC4335094
Genomic Map