Assay Details
| Gene Symbol: LOC4335530 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781114.2 | XP_015636600.1 | 4 - 5 | 462 | 114 | |
| GenBank mRNA | AK064621.1 | - | 4 - 5 | 387 | 114 | |
| KX434730.1 | - | 4 - 5 | 239 | 114 | ||
More Information
Set Membership: |
|

LOC4335530
Genomic Map