Assay Details
| Gene Symbol: LOC4335600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780778.2 | XP_015636264.1 | 3 - 4 | 769 | 77 | |
| XM_015780779.2 | XP_015636265.1 | 3 - 4 | 772 | 77 | ||
| GenBank mRNA | AK101615.1 | - | 3 - 4 | 625 | 77 | |
| Gene Symbol: LOC4350816 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK288325.1 | - | 2123 | 77 | ||
More Information
Set Membership: |
|

LOC4335600
Genomic Map