Assay Details
| Gene Symbol: LOC4335600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780778.2 | XP_015636264.1 | 6 - 7 | 1307 | 87 | |
| XM_015780779.2 | XP_015636265.1 | 6 - 7 | 1310 | 87 | ||
| GenBank mRNA | AK101615.1 | - | 6 - 7 | 1163 | 87 | |
| Gene Symbol: LOC4350816 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK288325.1 | - | 1585 | 87 | ||
More Information
Set Membership: |
|

LOC4335600
Genomic Map