Assay Details
| Gene Symbol: LOC4335896 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780377.1 | XP_015635863.1 | 9 - 10 | 1029 | 153 | |
| GenBank mRNA | AB114855.1 | - | 9 - 10 | 993 | 153 | |
| AK063934.1 | - | 9 - 10 | 1001 | 153 | ||
| AK103344.1 | - | 9 - 10 | 1002 | 153 | ||
More Information
Set Membership: |
|

LOC4335896
Genomic Map