Assay Details
| Gene Symbol: LOC4336076 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780639.2 | XP_015636125.1 | 2 - 3 | 369 | 141 | |
| GenBank mRNA | AB116721.1 | - | 2 - 3 | 295 | 141 | |
| AK107879.1 | - | 2 - 3 | 375 | 141 | ||
More Information
Set Membership: |
|

LOC4336076
Genomic Map