Assay Details
| Gene Symbol: LOC4336167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781800.2 | XP_015637286.1 | 10 - 11 | 1007 | 83 | |
| GenBank mRNA | AK106344.1 | - | 10 - 11 | 976 | 83 | |
| KF245924.1 | - | 10 - 11 | 996 | 83 | ||
More Information
Set Membership: |
|

LOC4336167
Genomic Map