Assay Details
| Gene Symbol: LOC4336167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781800.2 | XP_015637286.1 | 4 - 5 | 495 | 99 | |
| GenBank mRNA | AK106344.1 | - | 4 - 5 | 464 | 99 | |
| KF245924.1 | - | 4 - 5 | 484 | 99 | ||
More Information
Set Membership: |
|

LOC4336167
Genomic Map