Assay Details
| Gene Symbol: LOC4336257 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780844.1 | XP_015636330.1 | 4 - 5 | 197 | 81 | |
| XM_015780845.2 | XP_015636331.1 | 2 - 3 | 418 | 81 | ||
| XM_015780846.1 | XP_015636332.1 | 3 - 4 | 143 | 81 | ||
| XM_015780847.1 | XP_015636333.1 | 4 - 5 | 197 | 81 | ||
| GenBank mRNA | AK067292.1 | - | 2 - 3 | 399 | 81 | |
More Information
Set Membership: |
|

LOC4336257
Genomic Map