Assay Details
| Gene Symbol: LOC4336295 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779605.2 | XP_015635091.1 | 3 - 4 | 1461 | 107 | |
| GenBank mRNA | AK059661.1 | - | 3 - 4 | 1461 | 107 | |
| AK121011.1 | - | 3 - 4 | 1377 | 107 | ||
More Information
Set Membership: |
|

LOC4336295
Genomic Map