Assay Details
| Gene Symbol: LOC4336348 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781439.2 | XP_015636925.1 | 3 - 4 | 1123 | 57 | |
| GenBank mRNA | AK121383.1 | - | 3 - 4 | 1121 | 57 | |
More Information
Set Membership: |
|

LOC4336348
Genomic Map