Assay Details
| Gene Symbol: LOC4336402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778357.2 | XP_015633843.1 | 5 - 6 | 898 | 82 | |
| XM_015778358.2 | XP_015633844.1 | 5 - 6 | 893 | 82 | ||
| GenBank mRNA | AK059652.1 | - | 5 - 6 | 811 | 82 | |
| AK099280.1 | - | 5 - 6 | 809 | 82 | ||
More Information
Set Membership: |
|

LOC4336402
Genomic Map