Assay Details
| Gene Symbol: LOC4336402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778357.2 | XP_015633843.1 | 6 - 7 | 1026 | 107 | |
| XM_015778358.2 | XP_015633844.1 | 6 - 7 | 1021 | 107 | ||
| GenBank mRNA | AK059652.1 | - | 6 - 7 | 939 | 107 | |
| AK099280.1 | - | 6 - 7 | 937 | 107 | ||
More Information
Set Membership: |
|

LOC4336402
Genomic Map