Assay Details
| Gene Symbol: LOC4336402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778357.2 | XP_015633843.1 | 9 - 10 | 1262 | 106 | |
| XM_015778358.2 | XP_015633844.1 | 9 - 10 | 1257 | 106 | ||
| GenBank mRNA | AK059652.1 | - | 9 - 10 | 1175 | 106 | |
| AK099280.1 | - | 9 - 10 | 1173 | 106 | ||
More Information
Set Membership: |
|

LOC4336402
Genomic Map