Assay Details
| Gene Symbol: LOC107275790 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780952.2 | XP_015636438.1 | 4 - 5 | 1193 | 79 | |
| XM_015780953.2 | XP_015636439.1 | 4 - 5 | 1187 | 79 | ||
| XM_015780955.2 | XP_015636441.1 | 4 - 5 | 1280 | 79 | ||
| GenBank mRNA | AK059412.1 | - | 4 - 5 | 1121 | 79 | |
| AK103012.1 | - | 4 - 5 | 1283 | 79 | ||
More Information
Set Membership: |
|

LOC107275790
Genomic Map