Assay Details
| Gene Symbol: LOC4336584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780265.2 | XP_015635751.1 | 1 - 2 | 128 | 126 | |
| GenBank mRNA | AK101956.1 | - | 1 - 2 | 84 | 126 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015780265.2 transcripts.
Set Membership: |
|

LOC4336584
Genomic Map