Assay Details
| Gene Symbol: LOC4336584 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780265.2 | XP_015635751.1 | 5 - 6 | 750 | 100 | |
| XM_015780266.2 | XP_015635752.1 | 5 - 6 | 709 | 100 | ||
| GenBank mRNA | AK061922.1 | - | 4 - 5 | 528 | 100 | |
| AK101956.1 | - | 5 - 6 | 706 | 100 | ||
| EU325987.1 | - | 4 - 5 | 588 | 100 | ||
More Information
Set Membership: |
|

LOC4336584
Genomic Map