Assay Details
| Gene Symbol: LOC4336589 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780898.2 | XP_015636384.1 | 12 - 13 | 2108 | 104 | |
| GenBank mRNA | AK067475.1 | - | 12 - 13 | 1621 | 104 | |
More Information
Set Membership: |
|

LOC4336589
Genomic Map