Assay Details
| Gene Symbol: LOC4336845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780904.2 | XP_015636390.2 | 1 | 1446 | 79 | |
| GenBank mRNA | AK106907.1 | - | 1 | 1452 | 79 | |
More Information
Set Membership: |
|

LOC4336845
Genomic Map