Assay Details
| Gene Symbol: LOC4336870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779919.2 | XP_015635405.1 | 3 - 4 | 1262 | 123 | |
| XM_015779920.2 | XP_015635406.1 | 3 - 4 | 1259 | 123 | ||
| GenBank mRNA | AK103675.1 | - | 3 - 4 | 1239 | 123 | |
More Information
Set Membership: |
|

LOC4336870
Genomic Map