Assay Details
| Gene Symbol: LOC4336982 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780230.2 | XP_015635716.1 | 13 - 14 | 1110 | 94 | |
| XM_015780231.2 | XP_015635717.1 | 13 - 14 | 1200 | 94 | ||
| XM_015780232.2 | XP_015635718.1 | 13 - 14 | 1229 | 94 | ||
| GenBank mRNA | AK100259.1 | - | 13 - 14 | 1230 | 94 | |
More Information
Set Membership: |
|

LOC4336982
Genomic Map