Assay Details
| Gene Symbol: LOC4337007 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779488.2 | XP_015634974.1 | 6 - 7 | 749 | 145 | |
| GenBank mRNA | AK073351.1 | - | 6 - 7 | 758 | 145 | |
| BT131291.1 | - | 3 - 4 | 200 | 145 | ||
More Information
Set Membership: |
|

LOC4337007
Genomic Map