Assay Details
| Gene Symbol: LOC4337015 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779492.2 | XP_015634978.1 | 3 - 4 | 885 | 75 | |
| XM_015779494.2 | XP_015634980.1 | 2 - 3 | 521 | 75 | ||
| GenBank mRNA | AK106689.1 | - | 3 - 4 | 885 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015779492.2, XM_015779494.2 transcripts.
Set Membership: |
|

LOC4337015
Genomic Map