Assay Details
| Gene Symbol: LOC4337015 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779492.2 | XP_015634978.1 | 4 - 5 | 984 | 69 | |
| XM_015779494.2 | XP_015634980.1 | 3 - 4 | 620 | 69 | ||
| GenBank mRNA | AK106689.1 | - | 4 - 5 | 984 | 69 | |
More Information
Set Membership: |
|

LOC4337015
Genomic Map