Assay Details
| Gene Symbol: LOC4337074 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780852.2 | XP_015636338.1 | 10 - 11 | 2526 | 57 | |
| XM_015780853.2 | XP_015636339.1 | 11 - 12 | 2435 | 57 | ||
| XM_015780854.2 | XP_015636340.1 | 10 - 11 | 2080 | 57 | ||
| GenBank mRNA | AB101645.1 | - | 11 - 12 | 2232 | 57 | |
| AK065819.1 | - | 8 - 9 | 1529 | 57 | ||
| AK101563.1 | - | 7 - 8 | 1384 | 57 | ||
More Information
Set Membership: |
|

LOC4337074
Genomic Map