Assay Details
| Gene Symbol: LOC4337074 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780852.2 | XP_015636338.1 | 11 - 12 | 2955 | 85 | |
| XM_015780853.2 | XP_015636339.1 | 12 - 13 | 2864 | 85 | ||
| XM_015780854.2 | XP_015636340.1 | 11 - 12 | 2509 | 85 | ||
| GenBank mRNA | AB101645.1 | - | 12 - 13 | 2661 | 85 | |
| AK065819.1 | - | 9 - 10 | 1958 | 85 | ||
| AK101563.1 | - | 8 - 9 | 1813 | 85 | ||
More Information
Set Membership: |
|

LOC4337074
Genomic Map