Assay Details
| Gene Symbol: LOC4337074 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780852.2 | XP_015636338.1 | 1 - 2 | 638 | 61 | |
| XM_015780853.2 | XP_015636339.1 | 2 - 3 | 547 | 61 | ||
| GenBank mRNA | AB101645.1 | - | 2 - 3 | 344 | 61 | |
| AK065819.1 | - | 1 - 2 | 582 | 61 | ||
More Information
Set Membership: |
|

LOC4337074
Genomic Map