Assay Details
| Gene Symbol: LOC4337074 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780852.2 | XP_015636338.1 | 3 - 4 | 1034 | 103 | |
| XM_015780853.2 | XP_015636339.1 | 4 - 5 | 943 | 103 | ||
| XM_015780854.2 | XP_015636340.1 | 3 - 4 | 588 | 103 | ||
| GenBank mRNA | AB101645.1 | - | 4 - 5 | 740 | 103 | |
More Information
Set Membership: |
|

LOC4337074
Genomic Map