Assay Details
| Gene Symbol: LOC4337074 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780852.2 | XP_015636338.1 | 5 - 6 | 1680 | 73 | |
| XM_015780853.2 | XP_015636339.1 | 6 - 7 | 1589 | 73 | ||
| XM_015780854.2 | XP_015636340.1 | 5 - 6 | 1234 | 73 | ||
| GenBank mRNA | AB101645.1 | - | 6 - 7 | 1386 | 73 | |
| AK065819.1 | - | 3 - 4 | 683 | 73 | ||
| AK101563.1 | - | 2 - 3 | 538 | 73 | ||
More Information
Set Membership: |
|

LOC4337074
Genomic Map