Assay Details
| Gene Symbol: LOC4337654 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783843.2 | XP_015639329.1 | 5 - 6 | 1566 | 76 | |
| XM_015783844.2 | XP_015639330.1 | 4 - 5 | 1521 | 76 | ||
| GenBank mRNA | AY685117.1 | - | 5 - 6 | 1007 | 76 | |
More Information
Set Membership: |
|

LOC4337654
Genomic Map