Assay Details
| Gene Symbol: LOC4337654 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783843.2 | XP_015639329.1 | 7 - 8 | 1803 | 67 | |
| XM_015783844.2 | XP_015639330.1 | 6 - 7 | 1758 | 67 | ||
| GenBank mRNA | AY685117.1 | - | 7 - 8 | 1244 | 67 | |
More Information
Set Membership: |
|

LOC4337654
Genomic Map