Assay Details
| Gene Symbol: LOC4337226 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781373.2 | XP_015636859.1 | 1 | 967 | 91 | |
| XM_015781374.2 | XP_015636860.1 | 1 | 967 | 91 | ||
| XM_015781375.2 | XP_015636861.1 | 1 | 967 | 91 | ||
More Information
Set Membership: |
|

LOC4337226
Genomic Map