Assay Details
| Gene Symbol: LOC4337299 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780938.2 | XP_015636424.1 | 1 - 2 | 248 | 75 | |
| GenBank mRNA | AK065171.1 | - | 1 - 2 | 185 | 75 | |
| AK099504.1 | - | 1 - 2 | 186 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015780938.2 transcripts.
Set Membership: |
|

LOC4337299
Genomic Map