Assay Details
| Gene Symbol: LOC4337445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778474.2 | XP_015633960.1 | 1 - 2 | 505 | 65 | |
| XM_015778475.2 | XP_015633961.1 | 1 - 2 | 502 | 65 | ||
| GenBank mRNA | AK106642.1 | - | 1 - 2 | 465 | 65 | |
More Information
Set Membership: |
|

LOC4337445
Genomic Map