Assay Details
| Gene Symbol: LOC4337445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778474.2 | XP_015633960.1 | 3 - 4 | 680 | 108 | |
| XM_015778475.2 | XP_015633961.1 | 3 - 4 | 677 | 108 | ||
| XM_015778478.2 | XP_015633964.1 | 3 - 4 | 621 | 108 | ||
| XM_015778480.2 | XP_015633966.1 | 3 - 4 | 614 | 108 | ||
| GenBank mRNA | AK071538.1 | - | 3 - 4 | 583 | 108 | |
| AK105871.1 | - | 2 - 3 | 751 | 108 | ||
| AK106642.1 | - | 3 - 4 | 640 | 108 | ||
More Information
Set Membership: |
|

LOC4337445
Genomic Map