Assay Details
| Gene Symbol: LOC4337445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778475.2 | XP_015633961.1 | 7 - 8 | 1136 | 112 | |
| XM_015778480.2 | XP_015633966.1 | 7 - 8 | 1073 | 112 | ||
| GenBank mRNA | AK071538.1 | - | 7 - 8 | 1042 | 112 | |
| AK105871.1 | - | 5 - 6 | 1301 | 112 | ||
| AK106642.1 | - | 6 - 7 | 1214 | 112 | ||
More Information
Set Membership: |
|

LOC4337445
Genomic Map