Assay Details
| Gene Symbol: LOC4337845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784504.2 | XP_015639990.1 | 9 - 10 | 1907 | 83 | |
| GenBank mRNA | AB531488.1 | - | 9 - 10 | 1585 | 83 | |
| AK064328.1 | - | 2 - 3 | 527 | 83 | ||
More Information
Set Membership: |
|

LOC4337845
Genomic Map