Assay Details
| Gene Symbol: LOC4337925 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784942.2 | XP_015640428.1 | 1 - 2 | 210 | 141 | |
| XM_015784944.2 | XP_015640430.1 | 1 - 2 | 210 | 141 | ||
| GenBank mRNA | AK065618.1 | - | 1 - 2 | 119 | 141 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015784942.2, XM_015784944.2 transcripts.
Set Membership: |
|

LOC4337925
Genomic Map