Assay Details
| Gene Symbol: LOC4337925 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784942.2 | XP_015640428.1 | 7 - 8 | 1247 | 78 | |
| GenBank mRNA | AB060277.1 | - | 6 - 7 | 937 | 78 | |
| AK065618.1 | - | 7 - 8 | 1156 | 78 | ||
More Information
Set Membership: |
|

LOC4337925
Genomic Map