Assay Details
| Gene Symbol: LOC4338108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783450.2 | XP_015638936.1 | 2 - 3 | 280 | 108 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4338108| Gene Symbol: LOC4338108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783450.2 | XP_015638936.1 | 2 - 3 | 280 | 108 | |
Set Membership: |
|