Assay Details
| Gene Symbol: LOC4338204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782605.2 | XP_015638091.1 | 2 - 3 | 416 | 54 | |
| GenBank mRNA | AB091471.1 | - | 2 - 3 | 376 | 54 | |
| AK243380.1 | - | 2 - 3 | 342 | 54 | ||
More Information
Set Membership: |
|

LOC4338204
Genomic Map